4-Brom-N-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2800-1G HC-2800-5G
COBBHC-2800-1GEA 204 EUR
COBBHC-2800-1G COBBHC-2800-5G
4-Brom-N-methylanilin Hydrochlorid
4-Brom-N-methylanilin Hydrochlorid
Formel: C₇H₉BrClN
MDL: MFCD08456851
CAS-Nummer: 84250-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR