2,5-Dichlor-N-methylanilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-3070-5G
HC-3070-1G
COBBHC-3070-5GEA
886
EUR
COBBHC-3070-5G
COBBHC-3070-1G
2,5-Dichlor-N-methylanilin Hydrochlorid
2,5-Dichlor-N-methylanilin Hydrochlorid
Formel:
C₇H₈Cl₃N |
MDL:
MFCD12197277 CAS-Nummer: 1193389-58-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...