3-Chlor-N-methylanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2851-5G HC-2851-25G HC-2851-1G
COBBHC-2851-5GEA 49.7 EUR
COBBHC-2851-5G COBBHC-2851-25G COBBHC-2851-1G
3-Chlor-N-methylanilin Hydrochlorid
3-Chlor-N-methylanilin Hydrochlorid
Formel: C₇H₉Cl₂N
MDL: MFCD12756440
CAS-Nummer: 152428-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR