2-Fluor-N-methylanilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2849-5G
HC-2849-25G
COBBHC-2849-5GEA
86.4
EUR
COBBHC-2849-5G
COBBHC-2849-25G
2-Fluor-N-methylanilin Hydrochlorid
2-Fluor-N-methylanilin Hydrochlorid
Formel:
C₇H₉ClFN |
MDL:
MFCD12756438 CAS-Nummer: 1187386-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...