8-(Trifluormethyl)chinolin-6-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (8-(Trifluoromethyl)quinolin-6-yl)boronic acid

BB-5329-1G BB-5329-250M
COBBBB-5329-1GEA 1040 EUR
COBBBB-5329-1G COBBBB-5329-250M
8-(Trifluormethyl)chinolin-6-boronsäure
8-(Trifluormethyl)chinolin-6-boronsäure
Formel: C₁₀H₇BF₃NO₂
Molecular Weight: 240,9743 g/mol
MDL: MFCD11504852
CAS-Nummer: 1072951-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR