4-Pyrid-3-ylbenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 4-Pyridin-3-yl-benzoic acid

011855-500MG 011855-1G
FLUO011855-500MGEA 0 EUR
FLUO011855-500MG FLUO011855-1G
4-Pyrid-3-ylbenzoesäure
4-Pyrid-3-ylbenzoesäure
Formel: C₁₂H₉NO₂
Molecular Weight: 199,2054 g/mol
MDL: MFCD03426513
CAS-Nummer: 4385-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR