4-Pyrid-4-ylbenzoesäure

Lieferant: CHEMPUR

31-0001.500 31-0001.1
CHMP31-0001.500EA 0 EUR
CHMP31-0001.500 CHMP31-0001.1
4-Pyrid-4-ylbenzoesäure
4-Pyrid-4-ylbenzoesäure
Formel: C₁₂H₉NO₂
MDL: MFCD04114574
CAS-Nummer: 4385-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR