3-(4-Morpholino)propylisothiocyanat
Lieferant: FLUOROCHEM
009545-1G
009545-5G
009545-25G
FLUO009545-1GEA
34.1
EUR
FLUO009545-1G
FLUO009545-5G
FLUO009545-25G
3-(4-Morpholino)propylisothiocyanat
3-(4-Morpholino)propylisothiocyanat
Formel:
C₈H₁₄N₂OS Molecular Weight: 186.28 g/mol |
MDL:
MFCD00041222 CAS-Nummer: 32813-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...