1-(Cyanmethyl)pyridiniumchlorid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(Cyanomethyl)pyridin-1-ium chloride
A14327.09
A14327.18
A14327.09EA
38.1
EUR
A14327.09
A14327.18
1-(Cyanmethyl)pyridiniumchlorid ≥97%
1-(Cyanomethyl)pyridinium chloride
Purity: 97%
CAS No.: 17281-59-3
Molecular Formula: C7H7ClN2
Formula Weight: 154.60
Melting Point: 173-175°
Beilstein Reference: 3728392
UN No.: 3439
MDL No.: MFCD00031933
Formel:
C₇H₇ClN₂ Molecular Weight: 154,60 g/mol Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031933 CAS-Nummer: 17281-59-3 EINECS: 241-312-9 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...