4-(2,4-Dichlorphenyl)-3-thiosemicarbazid

Lieferant: BIONET RESEARCH

2P-045.500 2P-045.1 2P-045.5 2P-045.10
BIOT2P-045.500EA 0 EUR
BIOT2P-045.500 BIOT2P-045.1 BIOT2P-045.5 BIOT2P-045.10
4-(2,4-Dichlorphenyl)-3-thiosemicarbazid
4-(2,4-Dichlorphenyl)-3-thiosemicarbazid
Formel: C₇H₇Cl₂N₃S
Molecular Weight: 236.12 g/mol
MDL: MFCD00041272
CAS-Nummer: 13124-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR