4-(3,4-Dichlorphenyl)-3-thiosemicarbazid

Lieferant: BIONET RESEARCH

10T-0017.500 10T-0017.1 10T-0017.5 10T-0017.10
BIOT10T-0017.500EA 0 EUR
BIOT10T-0017.500 BIOT10T-0017.1 BIOT10T-0017.5 BIOT10T-0017.10
4-(3,4-Dichlorphenyl)-3-thiosemicarbazid
4-(3,4-Dichlorphenyl)-3-thiosemicarbazid
Formel: C₇H₇Cl₂N₃S
MDL: MFCD00041288
CAS-Nummer: 13124-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR