4-(2-Fluorphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

Synonyme: N-(2-Fluorophenyl)hydrazinecarbothioamide

009239-25G 009239-5G
FLUO009239-25GEA 453 EUR
FLUO009239-25G FLUO009239-5G
4-(2-Fluorphenyl)-3-thiosemicarbazid
4-(2-Fluorphenyl)-3-thiosemicarbazid
Formel: C₇H₈FN₃S
Molecular Weight: 185.22 g/mol
MDL: MFCD00041269
CAS-Nummer: 38985-72-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR