4-(4-Fluorphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

009241-10G 009241-50G
FLUO009241-10GEA 351 EUR
FLUO009241-10G FLUO009241-50G
4-(4-Fluorphenyl)-3-thiosemicarbazid
4-(4-Fluorphenyl)-3-thiosemicarbazid
Formel: C₇H₈FN₃S
Molecular Weight: 185,22 g/mol
MDL: MFCD00041294
CAS-Nummer: 330-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR