tert-Butylhydrazin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

194972-100G 194972-500G 194972-5G
SIAL194972-100GEA 81 EUR
SIAL194972-100G SIAL194972-500G SIAL194972-5G
tert-Butylhydrazin Hydrochlorid, Sigma-Aldrich®
tert-Butylhydrazin Hydrochlorid
Formel: C₄H₁₃ClN₂
Molecular Weight: 124,61 g/mol
MDL: MFCD00012947
CAS-Nummer: 7400-27-3
EINECS: 231-001-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR