4-Benzyl-3-thiosemicarbazid ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Benzylhydrazinecarbothioamide

L10920.03 L10920.06
L10920.03EA 45.1 EUR
L10920.03 L10920.06
4-Benzyl-3-thiosemicarbazid ≥98%
4-Benzyl-3-thiosemicarbazid

Purity: 98+% CAS No.: 13431-41-9 Molecular Formula: C8H11N3S Formula Weight: 181.26 Melting Point: 128-131° Beilstein Reference: 2096431 UN No.: 2811 MDL No.: MFCD00025145

Formel: C₈H₁₁N₃S
Molecular Weight: 181,26 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025145
CAS-Nummer: 13431-41-9
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR