Methyl-2-phenoxybenzoat ≥99%

Lieferant: Thermo Scientific

Synonyme: Methyl 2-phenoxybenzoate

L14584.09 L14584.18
L14584.09EA 27.4 EUR
L14584.09 L14584.18
Methyl-2-phenoxybenzoat ≥99%
Methyl-2-phenoxybenzoat

Purity: 99% CAS No.: 21905-56-6 Molecular Formula: C14H12O3 Formula Weight: 228.25 Refractive Index: 1.5760 Boiling Point: 130°/0.5mm MDL No.: MFCD03093064

Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
Siedepunkt: 130 °C (0,5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03093064
CAS-Nummer: 21905-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR