Methyl-3-phenoxybenzoat
Lieferant: CHEMPUR
Synonyme:
Methyl 3-phenoxybenzoate
SEW04723.250
SEW04723.1
CHMPSEW04723.250EA
0
EUR
CHMPSEW04723.250
MAYBSEW04723.1
Methyl-3-phenoxybenzoat
Methyl-3-phenoxybenzoat
Formel:
C₁₄H₁₂O₃ Molecular Weight: 228,2433 g/mol |
MDL:
MFCD00017192 CAS-Nummer: 50789-43-0 EINECS: 256-764-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...