2-(Methylsulfonyl)benzoesäure

Lieferant: BIONET RESEARCH

5L-603S.500 5L-603S.1 5L-603S.5 5L-603S.10
BIOT5L-603S.500EA 0 EUR
BIOT5L-603S.500 BIOT5L-603S.1 BIOT5L-603S.5 BIOT5L-603S.10
2-(Methylsulfonyl)benzoesäure
2-(Methylsulphonyl)benzoic acid
Formel: C₈H₈O₄S
Molecular Weight: 200,21 g/mol
MDL: MFCD00216492
CAS-Nummer: 33963-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR