4-(Dimethylamino)phenylisothiocyanat
Lieferant: FLUOROCHEM
005974-1G
005974-5G
FLUO005974-1GEA
31.1
EUR
FLUO005974-1G
FLUO005974-5G
4-(Dimethylamino)phenylisothiocyanat
4-(Dimethylamino)phenylisothiocyanat
Formel:
C₉H₁₀N₂S Molecular Weight: 178.25 g/mol |
MDL:
MFCD00041093 CAS-Nummer: 2131-64-8 EINECS: 218-360-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...