4-(Dimethylamino)phenylisothiocyanat
Lieferant: Thermo Scientific
S05025.1
S05025.10
S05025.50
MAYBS05025.1EA
0
EUR
MAYBS05025.1
MAYBS05025.10
MAYBS05025.50
4-(Dimethylamino)phenylisothiocyanat
4-(Dimethylamino)phenylisothiocyanat
Formel:
C₉H₁₀N₂S Molecular Weight: 178,25 g/mol |
MDL:
MFCD00041093 CAS-Nummer: 2131-64-8 EINECS: 218-360-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...