Methyl-2-methyl-3-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-methyl-3-nitrobenzoate

CA-4101-25G CA-4101-100G
COBBCA-4101-25GEA 50.9 EUR
COBBCA-4101-25G COBBCA-4101-100G
Methyl-2-methyl-3-nitrobenzoat
Methyl-2-methyl-3-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,17 g/mol
MDL: MFCD00082564
CAS-Nummer: 59382-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR