Methyl-3-methyl-2-nitrobenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 3-methyl-2-nitrobenzoate

11X-0063.500 11X-0063.1 11X-0063.5 11X-0063.10
BIOT11X-0063.500EA 0 EUR
BIOT11X-0063.500 BIOT11X-0063.1 BIOT11X-0063.5 BIOT11X-0063.10
Methyl-3-methyl-2-nitrobenzoat
Methyl-3-methyl-2-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
MDL: MFCD01003947
CAS-Nummer: 5471-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR