Methyl-3-methyl-4-nitrobenzoat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 3-methyl-4-nitrobenzoate

11X-0801.1 11X-0801.10 11X-0801.5 11X-0801.500
BIOT11X-0801.1EA 0 EUR
BIOT11X-0801.1 BIOT11X-0801.10 BIOT11X-0801.5 BIOT11X-0801.500
Methyl-3-methyl-4-nitrobenzoat
Methyl-3-methyl-4-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,18 g/mol
MDL: MFCD00085640
CAS-Nummer: 24078-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR