Methyl-3-methyl-4-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-methyl-4-nitrobenzoate

CA-4137-100G CA-4137-5G CA-4137-25G CA-4137-1G
COBBCA-4137-100GEA 127 EUR
COBBCA-4137-100G COBBCA-4137-5G COBBCA-4137-25G COBBCA-4137-1G
Methyl-3-methyl-4-nitrobenzoat
Methyl-3-methyl-4-nitrobenzoat
Formel: C₉H₉NO₄
Molecular Weight: 195,18 g/mol
MDL: MFCD00085640
CAS-Nummer: 24078-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR