4-(Benzyloxy)phenylisothiocyanat

Lieferant: FLUOROCHEM

008744-5G 008744-1G
FLUO008744-5GEA 227 EUR
FLUO008744-5G FLUO008744-1G
4-(Benzyloxy)phenylisothiocyanat
4-(Benzyloxy)phenylisothiocyanat
Formel: C₁₄H₁₁NOS
Molecular Weight: 241.31 g/mol
MDL: MFCD00041095
CAS-Nummer: 139768-71-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR