7-Chlor-4-chinolinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
7-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline
PN-8723-1G
PN-8723-250M
COBBPN-8723-1GEA
288
EUR
COBBPN-8723-1G
COBBPN-8723-250M
7-Chlor-4-chinolinboronsäurepinakolester
7-Chlor-4-chinolinboronsäurepinakolester
Formel:
C₁₅H₁₇BClNO₂ Molecular Weight: 289.56 g/mol |
MDL:
MFCD07783020 CAS-Nummer: 871125-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...