2-Chlorchinolin-3-boronsäure
Lieferant: COMBI-BLOCKS
BB-2262-5G
BB-2262-1G
COBBBB-2262-5GEA
229
EUR
COBBBB-2262-5G
COBBBB-2262-1G
2-Chlorchinolin-3-boronsäure
2-Chlorchinolin-3-boronsäure
Formel:
C₉H₇BClNO₂ Molecular Weight: 207,4214 g/mol |
MDL:
MFCD01114675 CAS-Nummer: 128676-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...