3-Methyl-2-butylisothiocyanat

Lieferant: FLUOROCHEM

009527-1G 009527-5G
FLUO009527-1GEA 55.4 EUR
FLUO009527-1G FLUO009527-5G
3-Methyl-2-butylisothiocyanat
3-Methyl-2-butylisothiocyanat
Formel: C₆H₁₁NS
MDL: MFCD00060388
CAS-Nummer: 201224-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR