4-tert-Butylphenylisothiocyanat

Lieferant: Thermo Scientific

Synonyme: 4-tert-Butylphenylisothiocyanate

TL00746DA TL00746EA TL00746.50
MAYBTL00746DAEA 0 EUR
MAYBTL00746DA MAYBTL00746EA MAYBTL00746.50
4-tert-Butylphenylisothiocyanat
4-tert-Butylphenylisothiocyanat
Formel: C₁₁H₁₃NS
Molecular Weight: 191,2926 g/mol
MDL: MFCD00022059
CAS-Nummer: 19241-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR