Methyl-3-isothiocyanatobenzoat

Lieferant: FLUOROCHEM

Synonyme: 3-(Methoxycarbonyl)phenyl isothiocyanate

009504-1G 009504-5G
FLUO009504-1GEA 52.5 EUR
FLUO009504-1G FLUO009504-5G
Methyl-3-isothiocyanatobenzoat
Methyl-3-isothiocyanatobenzoat
Formel: C₉H₇NO₂S
MDL: MFCD00041080
CAS-Nummer: 3125-66-4
EINECS: 240-164-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR