Ethyl-4-isothiocyanatobenzoat
Lieferant: FLUOROCHEM
Synonyme:
4-(Ethoxycarbonyl)phenyl isothiocyanate
008940-5G
008940-25G
FLUO008940-5GEA
57.9
EUR
FLUO008940-5G
FLUO008940-25G
Ethyl-4-isothiocyanatobenzoat
Ethyl-4-isothiocyanatobenzoat
Formel:
C₁₀H₉NO₂S Molecular Weight: 207,25 g/mol |
MDL:
MFCD00022058 CAS-Nummer: 1205-06-7 EINECS: 214-880-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...