Methyl-4-isothiocyanatobenzoat ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(Methoxycarbonyl)phenyl isothiocyanate

L10474.03 L10474.06
L10474.03EA 59.3 EUR
L10474.03 L10474.06
Methyl-4-isothiocyanatobenzoat ≥98%
Methyl-4-isothiocyanatobenzoat

Purity: 98+% CAS No.: 3662-78-0 Molecular Formula: C9H7NO2S Formula Weight: 193.23 Odour: Irritating Melting Point: 46-50° Beilstein Reference: 2100504 UN No.: 2923 MDL No.: MFCD00041099

Formel: C₉H₇NO₂S
Molecular Weight: 193,23 g/mol
Schmelzpunkt: 46…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041099
CAS-Nummer: 3662-78-0
EINECS: 240-164-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR