Methyl-4-isothiocyanatobenzoat

Lieferant: FLUOROCHEM

Synonyme: 4-(Methoxycarbonyl)phenyl isothiocyanate

009505-1G 009505-5G
FLUO009505-1GEA 119 EUR
FLUO009505-1G FLUO009505-5G
Methyl-4-isothiocyanatobenzoat
Methyl-4-isothiocyanatobenzoat
Formel: C₉H₇NO₂S
Molecular Weight: 193,23 g/mol
MDL: MFCD00041099
CAS-Nummer: 3662-78-0
EINECS: 240-164-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR