4-Methoxy-2-nitrophenylisothiocyanat
Lieferant: FLUOROCHEM
009512-1G
009512-5G
FLUO009512-1GEA
88.1
EUR
FLUO009512-1G
FLUO009512-5G
4-Methoxy-2-nitrophenylisothiocyanat
4-Methoxy-2-nitrophenylisothiocyanat
Formel:
C₈H₆N₂O₃S Molecular Weight: 210,21 g/mol |
MDL:
MFCD00041380 CAS-Nummer: 23165-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...