2-(3,4-Dimethoxyphenyl)ethylisothiocyanat

Lieferant: Thermo Scientific

TL00669DA TL00669EA TL00669.50
MAYBTL00669DAEA 0 EUR
MAYBTL00669DA MAYBTL00669EA MAYBTL00669.50
2-(3,4-Dimethoxyphenyl)ethylisothiocyanat
2-(3,4-Dimethoxyphenyl)ethylisothiocyanat
Formel: C₁₁H₁₃NO₂S
Molecular Weight: 223,30 g/mol
MDL: MFCD00041366
CAS-Nummer: 21714-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR