4-Mercaptobenzoesäure

Lieferant: FLUOROCHEM

Synonyme: 4-Thiobenzoic acid

013962-1G 013962-5G
FLUO013962-1GEA 60.9 EUR
FLUO013962-1G FLUO013962-5G
4-Mercaptobenzoesäure
4-Mercaptobenzoic acid
Formel: C₇H₆O₂S
MDL: MFCD00016617
CAS-Nummer: 1074-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR