4-Fluor-2-iodbenzoesäure ≥97%

Lieferant: Thermo Scientific
Gefahr

H55531.03 H55531.06
H55531.03EA 57.1 EUR
H55531.03 H55531.06
4-Fluor-2-iodbenzoesäure ≥97%
4-Fluoro-2-iodobenzoic acid

Purity: 97% CAS No.: 56096-89-0 Molecular Formula: C7H4FIO2 Formula Weight: 266.01 Melting Point: 143-147° UN No.: 2811 MDL No.: MFCD04108278

Formel: C₇H₄FIO₂
Molecular Weight: 266,01 g/mol
Schmelzpunkt: 143…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04108278
CAS-Nummer: 56096-89-0
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR