8-Fluorchinolin-7-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(8-Fluoroquinolin-7-yl)boronic acid
BB-5805-1G
BB-5805-250M
COBBBB-5805-1GEA
864
EUR
COBBBB-5805-1G
COBBBB-5805-250M
8-Fluorchinolin-7-boronsäure
8-Fluorchinolin-7-boronsäure
Formel:
C₉H₇BFNO₂ Molecular Weight: 190,970 g/mol |
MDL:
MFCD12025987 CAS-Nummer: 1217500-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...