2-Methoxy-4-nitrophenylisothiocyanat

Lieferant: FLUOROCHEM

009510-1G 009510-5G
FLUO009510-1GEA 48.8 EUR
FLUO009510-1G FLUO009510-5G
2-Methoxy-4-nitrophenylisothiocyanat
2-Methoxy-4-nitrophenylisothiocyanat
Formel: C₈H₆N₂O₃S
Schmelzpunkt: 107…109 °C
MDL: MFCD00041379
CAS-Nummer: 190774-55-1
UN: 2923
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR