2-Methoxy-5-methylphenylisothiocyanat

Lieferant: FLUOROCHEM

009508-5G 009508-25G
FLUO009508-5GEA 0 EUR
FLUO009508-5G FLUO009508-25G
2-Methoxy-5-methylphenylisothiocyanat
2-Methoxy-5-methylphenylisothiocyanat
Formel: C₉H₉NOS
Molecular Weight: 179,24 g/mol
Siedepunkt: 129…130 °C (0,5 mmHg)
Dichte: 1,146 g/cm³ (20 °C)
MDL: MFCD00041083
CAS-Nummer: 190774-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR