SRK® Short Swab Rinse Kits
Lieferant: Copan
922C 926C 929C 947C 949C 924C
710-0477EA 90.2 EUR
710-0477 710-0478 710-0932 710-0933 COPA949C COPA924C
SRK® Short Swab Rinse Kits
Tupfer Tupfer im Set mit Spüllösung
Swab rinse kits (SRK®) for microbiological examination of surfaces.
  • Isotonic salt solution to neutralise and deactivate disinfectants and sanitising agents
  • Rinse solution to moisten the sterile swab before sampling and transport it after collection
  • Swab shafts (rayon tip) are attached to the tube caps
Suitable for food and industrial applications.
Lieferumfang: Jede Schachtel enthält 50 Röhrchen mit Schraubverschluss.