Rain trousers, Murray 699Z
Lieferant: SIOEN PERSONAL PROTECTIVE CLOT
699Z/A41/1XL 699Z/A41/2XL 699Z/A41/3XL 699Z/A41/L 699Z/A41/M 699Z/A41/S 699Z/B75/1XL 699Z/B75/2XL 699Z/B75/3XL 699Z/B75/L 699Z/B75/M 699Z/B75/S
SIOE699Z/A41/1XLEA 42.3 EUR
SIOE699Z/A41/1XL SIOE699Z/A41/2XL SIOE699Z/A41/3XL SIOE699Z/A41/L SIOE699Z/A41/M SIOE699Z/A41/S SIOE699Z/B75/1XL SIOE699Z/B75/2XL SIOE699Z/B75/3XL SIOE699Z/B75/L SIOE699Z/B75/M SIOE699Z/B75/S
Rain trousers, Murray 699Z
Arbeitskleidung
Highly breathable rain trousers made of Siopor® Regular fabric consisting of 100% polyester fabric (plain weave) with water-repellent PU coating. With fixed mesh lining and taped seams.
  • Supple and lightweight
  • Legs adjustable by press studs
  • Elasticated drawstring in waist
Weight: 155 g/m²
Standards: EN 343 (3-3)