N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
3-(N-Benzylsulphamoyl)-5-bromobenzotrifluoride
HC-6223-5G
HC-6223-1G
COBBHC-6223-5GEA
595
EUR
COBBHC-6223-5G
COBBHC-6223-1G
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
N-Benzyl-3-brom-5-(trifluormethyl)benzolsulfonamid
Formel:
C₁₄H₁₁BrF₃NO₂S |
MDL:
MFCD09800987 CAS-Nummer: 951885-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...