2-Brom-3-nitrotoluol

Lieferant: COMBI-BLOCKS

OS-7714-25G OS-7714-5G OS-7714-1G
COBBOS-7714-25GEA 166 EUR
COBBOS-7714-25G COBBOS-7714-5G COBBOS-7714-1G
2-Brom-3-nitrotoluol
2-Brom-3-nitrotoluol
Formel: C₇H₆BrNO₂
Molecular Weight: 216.0300 g/mol
MDL: MFCD00134555
CAS-Nummer: 41085-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR