2-Chlor-(5-ethoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-(ethoxycarbonyl)phenylboronic acid, (2-Chloro-5-(ethoxycarbonyl)phenyl)boronic acid

BB-3529-5G BB-3529-1G
COBBBB-3529-5GEA 692 EUR
COBBBB-3529-5G COBBBB-3529-1G
2-Chlor-(5-ethoxycarbonyl)phenylboronsäure
2-Chlor-(5-ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₀BClO₄
Molecular Weight: 228.4373 g/mol
MDL: MFCD08689529
CAS-Nummer: 913835-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR