2-Chlor-(5-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-(methoxycarbonyl)phenylboronic acid, (2-Chloro-5-(methoxycarbonyl)phenyl)boronic acid, 2-Chloro-5-(methoxycarbonyl)benzeneboronic acid

BB-3528-5G BB-3528-1G
COBBBB-3528-5GEA 834 EUR
COBBBB-3528-5G COBBBB-3528-1G
2-Chlor-(5-methoxycarbonyl)phenylboronsäure
2-Chlor-(5-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BClO₄
Molecular Weight: 214.41 g/mol
MDL: MFCD08689528
CAS-Nummer: 913835-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR