3-Carboxy-4-chlorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Borono-2-chlorobenzoic acid

BB-2798-1G BB-2798-5G
COBBBB-2798-1GEA 48.6 EUR
COBBBB-2798-1G COBBBB-2798-5G
3-Carboxy-4-chlorphenylboronsäure
3-Carboxy-4-chlorphenylboronsäure
Formel: C₇H₆BClO₄
Molecular Weight: 200.3800 g/mol
MDL: MFCD06801689
CAS-Nummer: 913835-32-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR