5-Carboxy-2-chlor-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3285-1G BB-3285-250M
COBBBB-3285-1GEA 1110 EUR
COBBBB-3285-1G COBBBB-3285-250M
5-Carboxy-2-chlor-4-fluorphenylboronsäure
5-Carboxy-2-chlor-4-fluorphenylboronsäure
Formel: C₇H₅BClFO₄
MDL: MFCD09878325
CAS-Nummer: 957066-06-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR