5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-(Dimethylcarbamoyl)-2-fluorobenzeneboronic acid, (5-(Dimethylcarbamoyl)-2-fluorophenyl)boronic acid, 5-Dimethylcarbamoyl-2-fluorobenzeneboronic acid
BB-3341-5G
BB-3341-1G
COBBBB-3341-5GEA
0
EUR
COBBBB-3341-5G
COBBBB-3341-1G
5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
5-(Dimethylcarbamoyl)-2-fluorphenylboronsäure
Formel:
C₉H₁₁BFNO₃ |
MDL:
MFCD08235102 CAS-Nummer: 874289-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...