4-Benzylcarbamoyl-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(Benzylcarbamoyl)-3-fluorophenyl)boronic acid, 4-(Benzylcarbamoyl)-3-fluorophenylboronic acid

BB-3315-5G BB-3315-1G
COBBBB-3315-5GEA 1490 EUR
COBBBB-3315-5G COBBBB-3315-1G
4-Benzylcarbamoyl-3-fluorphenylboronsäure
4-Benzylcarbamoyl-3-fluorphenylboronsäure
Formel: C₁₄H₁₃BFNO₃
Molecular Weight: 273,0700 g/mol
MDL: MFCD08689501
CAS-Nummer: 874289-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR